Learn More
ABclonal Technology MIIP Rabbit pAb
SDP
Supplier:  ABclonal Technology A144171000UL
Encompass_Preferred
This gene encodes a protein that interacts with the oncogene protein insulin-like growth factor binding protein 2 and may function as an inhibitor of cell migration and invasion. This protein also interacts with the cell division protein 20 and may be involved in regulating mitotic progression. This protein may function as a tumor suppressor by inhibiting the growth or certain cancers.
Catalog No. 50-001-08339