Learn More
Medchemexpress LLC HY-112218 10mg Medchemexpress, MIK665 CAS:1799631-75-6 Purity:>98%
SDP
Catalog No. 501969064
Encompass_Preferred
Medchemexpress, HY-112218 10mg MIK665 CAS:1799631-75-6 MIK665 (S-64315) is a special Mcl-1 inhibitor extracted from patent WO2016207225A1, compound Preparation 13, has an IC50 of 1.81 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9064 Supplier Medchemexpress LLC Supplier No. HY11221810MG
Medchemexpress, HY-112218 10mg MIK665 CAS:1799631-75-6 MIK665 (S-64315) is a special Mcl-1 inhibitor extracted from patent WO2016207225A1, compound Preparation 13, has an IC50 of 1.81 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.