Learn More
Selleck Chemical LLC MIK665 S64315-1mg
SDP
Catalog No. 502389666
Encompass_Preferred
MIK665 (S64315) is an inhibitor of induced myeloid leukemia cell differentiation protein Mcl-1 with Ki value of 1.2 nM and has potential pro-apoptotic and antineoplastic activities.
Catalog No. 50-238-9666 Supplier Selleck Chemical LLC Supplier No. S88361MG
MIK665 (S64315) is an inhibitor of induced myeloid leukemia cell differentiation protein Mcl-1 with Ki value of 1.2 nM and has potential pro-apoptotic and antineoplastic activities.