Learn More
Hettich Centrifuges MIKRO 185, (12) 1.5-2.0mL,Package
SDP
Catalog No. 502537696 Shop All Hettich Centrifuges Products
Encompass_Preferred
Hettich MIKRO 185 MICROLITER TUBE PACKAGE 12, (1) Centrifuge, (1) Fixed-angle Rotor, (12) Adapters 1.5mL, (1) Lid, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years
Catalog No. 50-253-7696 Supplier Hettich Centrifuges Supplier No. 185MICRO12
Add to Cart
Add to Cart
Hettich MIKRO 185 MICROLITER TUBE PACKAGE 12, (1) Centrifuge, (1) Fixed-angle Rotor, (12) Adapters 1.5mL, (1) Lid, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years