Learn More
Hettich Centrifuges MIKRO 185,(18) SpinCol,Bio,Package
SDP
Catalog No. 502537692 Shop All Hettich Centrifuges Products
Encompass_Preferred
Hettich MIKRO 185 Spin Column Package 18, (1) Centrifuge, (1) Fixed-angle Rotor, (18) Adapters 0.5mL, (1) Bio-containment Lid, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years
Catalog No. 50-253-7692 Supplier Hettich Centrifuges Supplier No. 185MICROSCBC
Hettich MIKRO 185 Spin Column Package 18, (1) Centrifuge, (1) Fixed-angle Rotor, (18) Adapters 0.5mL, (1) Bio-containment Lid, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years