Learn More
Hettich Centrifuges MIKRO 200, (20) Cryo, Bio, Package
SDP
Catalog No. 502537688 Shop All Hettich Centrifuges Products
Encompass_Preferred
Hettich MIKRO 200 Cryo Tube Package, (1) Centrifuge, (1) Fixed-angle Rotor, (1) Biocontainment lid, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years
Catalog No. 50-253-7688 Supplier Hettich Centrifuges Supplier No. 200CRYOBC
Add to Cart
Add to Cart
Request a Quote
Hettich MIKRO 200 Cryo Tube Package, (1) Centrifuge, (1) Fixed-angle Rotor, (1) Biocontainment lid, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years