Learn More
Hettich Centrifuges MIKRO 200,(24) 1.5-2.0mL,Package
SDP
Catalog No. 502537686 Shop All Hettich Centrifuges Products
Encompass_Preferred
Hettich MIKRO 200 MICROLITER TUBE PACKAGE 24 BIO-CONTAINMENT, (1) Centrifuge, (1) Fixed-angle Rotor, (24) Adapters 1.5mL, (1) Bio-containment Lid, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years
Catalog No. 50-253-7686 Supplier Hettich Centrifuges Supplier No. 200MICRO24
Hettich MIKRO 200 MICROLITER TUBE PACKAGE 24 BIO-CONTAINMENT, (1) Centrifuge, (1) Fixed-angle Rotor, (24) Adapters 1.5mL, (1) Bio-containment Lid, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years