Learn More
Hettich Centrifuges MIKRO 200R,(24) SpinColumn,Package
SDP
Catalog No. 502537677 Shop All Hettich Centrifuges Products
Encompass_Preferred
Hettich MIKRO 200R SPIN COLUMN KIT PACKAGE BIO-CONTAINMENT, (1) Centrifuge, (1) Fixed-angle Rotor, (1) Bio-containment Lid, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years
Catalog No. 50-253-7677 Supplier Hettich Centrifuges Supplier No. 200RMICROSCBC
Add to Cart
Add to Cart
Request a Quote
Hettich MIKRO 200R SPIN COLUMN KIT PACKAGE BIO-CONTAINMENT, (1) Centrifuge, (1) Fixed-angle Rotor, (1) Bio-containment Lid, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years