Learn More
Hettich Centrifuges MIKRO 200R,(30) 1.5-2.0mL,Package
SDP
Catalog No. 502537678 Shop All Hettich Centrifuges Products
Encompass_Preferred
Hettich MIKRO 200R MICROLITER TUBE PACKAGE 30 BIO-CONTAINMENT, (1) Centrifuge, (1) Fixed-angle Rotor, (30) Adapters 1.5mL, (1) Bio-containment Lid, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years
Catalog No. 50-253-7678 Supplier Hettich Centrifuges Supplier No. 200RMICRO30BC
Add to Cart
Add to Cart
Request a Quote
Hettich MIKRO 200R MICROLITER TUBE PACKAGE 30 BIO-CONTAINMENT, (1) Centrifuge, (1) Fixed-angle Rotor, (30) Adapters 1.5mL, (1) Bio-containment Lid, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years