Learn More
Hettich Centrifuges MIKRO 200R, 4 x 8 PCR strips
SDP
Catalog No. 502537676 Shop All Hettich Centrifuges Products
Encompass_Preferred
Hettich MIKRO 200R MICROLITER TUBE PACKAGE STRIP, (1) Centrifuge, (1) Fixed-angle Rotor, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years
Catalog No. 50-253-7676 Supplier Hettich Centrifuges Supplier No. 200RMICROSTRIP
Add to Cart
Add to Cart
Request a Quote
Hettich MIKRO 200R MICROLITER TUBE PACKAGE STRIP, (1) Centrifuge, (1) Fixed-angle Rotor, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years