Learn More
Hettich Centrifuges MIKRO 220R, (30) 1.5-2.0mL,Package
SDP
Catalog No. 502537661 Shop All Hettich Centrifuges Products
Encompass_Preferred
Hettich MIKRO 220R MICROLITER TUBE PACKAGE 30, (1) Centrifuge, (1) Fixed-Angle Rotor, (30) Adapters 1.5mL, (1) Biocontainment lid, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years
Catalog No. 50-253-7661 Supplier Hettich Centrifuges Supplier No. 220RMICRO30
Add to Cart
Add to Cart
Request a Quote
Hettich MIKRO 220R MICROLITER TUBE PACKAGE 30, (1) Centrifuge, (1) Fixed-Angle Rotor, (30) Adapters 1.5mL, (1) Biocontainment lid, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years