Learn More
Hettich Centrifuges MIKRO 220R,(6) 15,(3) 50,Package
SDP
Catalog No. 502537665 Shop All Hettich Centrifuges Products
Encompass_Preferred
Hettich MIKRO 220R MICROLITER CELL CULTURE PACKAGE, (1) Centrifuge, (1) Fixed-angle Rotor, (6) Adapters 15mL, (3) Adapters 50ml, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years
Catalog No. 50-253-7665 Supplier Hettich Centrifuges Supplier No. 220RCELLCULTURE
Add to Cart
Add to Cart
Request a Quote
Hettich MIKRO 220R MICROLITER CELL CULTURE PACKAGE, (1) Centrifuge, (1) Fixed-angle Rotor, (6) Adapters 15mL, (3) Adapters 50ml, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years