Learn More
Hettich Centrifuges MIKRO 220R, CryoTubes, Bio,Package
SDP
Catalog No. 502537663 Shop All Hettich Centrifuges Products
Encompass_Preferred
Hettich MIKRO R 220 Cryo Tube Package, (1) Centrifuge, (1) Fixed-angle Rotor, (1) Biocontainment Lid, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years
Catalog No. 50-253-7663 Supplier Hettich Centrifuges Supplier No. 220RCRYOBC
Add to Cart
Add to Cart
Request a Quote
Hettich MIKRO R 220 Cryo Tube Package, (1) Centrifuge, (1) Fixed-angle Rotor, (1) Biocontainment Lid, Power: 100-120 V 1~/50-60Hz, Warranty: Five (5) years