Learn More
Electron Microscopy Sciences Mikrostik™ Adhesive, Non-Conductive 14 ML
SDP
Supplier:  Electron Microscopy Sciences 1264601
Encompass_Preferred
Fast drying, ultrathin clear adhesive suitable for mounting small particles which can be submerged in other adhesives. It can be diluted with methyl ethyl ketone. Quick-drying. Comes in a bottle with an applicator brush. 14 ml.
Catalog No. 50-192-6787