Learn More
Medchemexpress LLC Milademetan | 1398568-47-2 | MFCD31692325 | 98.9% | 618.53 g/mol | C30H34Cl2FN5O4 | 1 MG
SDP
Catalog No. 5000212661
Encompass_Preferred
Milademetan is a potent, orally active small-molecule inhibitor of the MDM2-p53 interaction used in preclinical oncology research. It restores p53 activity and induces G1 cell-cycle arrest, senescence, and apoptosis in p53 wild-type cancer models.
  • Orally active MDM2-p53 inhibitor suitable for in vitro and in vivo research.
  • Applicable to acute myeloid leukemia and various solid tumor studies.
  • Induces G1 cell-cycle arrest, senescence, and apoptosis in p53 wild-type models.
  • High reported purity for research use (≈98.9%).
  • Well-characterized molecular properties (molecular weight 618.53 g/mol).
Catalog No. 50-002-12661 Supplier Medchemexpress LLC Supplier No. HY1012661MG
Milademetan is a potent, orally active small-molecule inhibitor of the MDM2-p53 interaction used in preclinical oncology research. It restores p53 activity and induces G1 cell-cycle arrest, senescence, and apoptosis in p53 wild-type cancer models.
  • Orally active MDM2-p53 inhibitor suitable for in vitro and in vivo research.
  • Applicable to acute myeloid leukemia and various solid tumor studies.
  • Induces G1 cell-cycle arrest, senescence, and apoptosis in p53 wild-type models.
  • High reported purity for research use (≈98.9%).
  • Well-characterized molecular properties (molecular weight 618.53 g/mol).