Learn More
TARGETMOL CHEMICALS INC MILADEMETAN 5MG
SDP
Catalog No. 502581487
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Milademetan (DS-3032) is an orally active and potent MDM2 inhibitor with antitumor activity that induces G1 cell cycle arrest senescence and apoptosis.Milademetan is used in the study of acute myeloid leukemia and solid tumors. purity: 100%
Catalog No. 50-258-1487 Supplier TARGETMOL CHEMICALS INC Supplier No. T120405MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Milademetan (DS-3032) is an orally active and potent MDM2 inhibitor with antitumor activity that induces G1 cell cycle arrest senescence and apoptosis.Milademetan is used in the study of acute myeloid leukemia and solid tumors. purity: 100%