Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Milbemycin A3 | 51596-10-2 | MFCD04972569 | 99.5% | 528.68 g/mol | C31H44O7 | 5 MG
SDP

Catalog No. 5000262880
Encompass_Preferred

Milbemycin A3 is an insecticidal antibiotic exhibiting acaricidal and nematocidal activity, supplied as a solid for research use. It is used in laboratory studies of parasitic control and insecticidal effects.

  • Has acaricidal and nematocidal activity.
  • Supplied as a solid, white to light yellow powder.
  • Chemical formula C31H44O7; molecular weight 528.68 g/mol.
  • CAS number 51596-10-2.
  • Reported purity varies by supplier (reported ranges include ≥85% to ≈99.5%).
  • Intended for research use only.

Catalog No. 50-002-62880 Supplier Medchemexpress LLC Supplier No. HY1304235MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Milbemycin A3 is an insecticidal antibiotic exhibiting acaricidal and nematocidal activity, supplied as a solid for research use. It is used in laboratory studies of parasitic control and insecticidal effects.

  • Has acaricidal and nematocidal activity.
  • Supplied as a solid, white to light yellow powder.
  • Chemical formula C31H44O7; molecular weight 528.68 g/mol.
  • CAS number 51596-10-2.
  • Reported purity varies by supplier (reported ranges include ≥85% to ≈99.5%).
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.