Learn More
Cayman Chemical MIlbemycIn oxIme 100mg
SDP
Catalog No. 502979934
Encompass_Preferred
A semisynthetic macrocyclic lactone prepared by the oxidation and oximation of an 80:20 ratio of milbemycin A3 and A4; nematocide and insecticide activity via the potentiation of glutamate and GABA-gated chloride-channel opening
Catalog No. 50-297-9934 Supplier Cayman Chemical Supplier No. 17164100 MG
A semisynthetic macrocyclic lactone prepared by the oxidation and oximation of an 80:20 ratio of milbemycin A3 and A4; nematocide and insecticide activity via the potentiation of glutamate and GABA-gated chloride-channel opening