Learn More
Medchemexpress LLC Milbemycin oxime | 129496-10-2 | 99.5% | 1097.38 | 50 MG
SDP
Catalog No. 5000366143
Encompass_Preferred
Milbemycin oxime is an orally active macrolide that exhibits broad-spectrum antiparasitic activity. It is comprised of oxime derivatives corresponding to milbemycin A4 and A3. This compound functions by binding to glutamate-gated chloride channels, thereby demonstrating inhibitory effects against various internal parasites, including intestinal nematodes and lung/heart worms.
  • Exhibits broad-spectrum action against parasites.
  • Active when administered orally.
  • Targets specific ion channels in parasites.
  • Effective against various types of worms.
Catalog No. 50-003-66143 Supplier Medchemexpress LLC Supplier No. 50MGHYB0778
Milbemycin oxime is an orally active macrolide that exhibits broad-spectrum antiparasitic activity. It is comprised of oxime derivatives corresponding to milbemycin A4 and A3. This compound functions by binding to glutamate-gated chloride channels, thereby demonstrating inhibitory effects against various internal parasites, including intestinal nematodes and lung/heart worms.
  • Exhibits broad-spectrum action against parasites.
  • Active when administered orally.
  • Targets specific ion channels in parasites.
  • Effective against various types of worms.