Learn More
Medchemexpress LLC Milbemycin oxime | 129496-10-2 | 99.5% | 5 MG
SDP
Catalog No. 5000409277
Encompass_Preferred
Milbemycin oxime is an orally active macrolide with broad-spectrum antiparasitic activity. It is a mixture of oximes consisting of oxime derivatives corresponding to milbemycin A4 and A3. This compound binds to glutamate-gated chloride channels and demonstrates inhibitory potency against intestinal nematodes and lung/heart worms.
  • Orally active macrolide
  • Offers broad-spectrum antiparasitic activity
  • Binds to glutamate-gated chloride channels
  • Inhibits intestinal nematodes
  • Inhibits lung and heart worms
Catalog No. 50-004-09277 Supplier Medchemexpress LLC Supplier No. HYB07785MG
Milbemycin oxime is an orally active macrolide with broad-spectrum antiparasitic activity. It is a mixture of oximes consisting of oxime derivatives corresponding to milbemycin A4 and A3. This compound binds to glutamate-gated chloride channels and demonstrates inhibitory potency against intestinal nematodes and lung/heart worms.
  • Orally active macrolide
  • Offers broad-spectrum antiparasitic activity
  • Binds to glutamate-gated chloride channels
  • Inhibits intestinal nematodes
  • Inhibits lung and heart worms