Learn More
Selleck Chemical LLC Milbemycin Oxime S5055-25mg
SDP
Catalog No. 503112236
Encompass_Preferred
Milbemycin Oxime is a broad spectrum anti-parasitic drug that is active against worms (anthelmintic) and mites (miticide)
Catalog No. 50-311-2236 Supplier Selleck Chemical LLC Supplier No. S505525MG
Milbemycin Oxime is a broad spectrum anti-parasitic drug that is active against worms (anthelmintic) and mites (miticide)