Learn More
Cayman Chemical Milbemycin oxime, Purity: ≥95%, CAS: 129496-10-2, 50 mg
SDP
Catalog No. 502979917
Encompass_Preferred
A semisynthetic macrocyclic lactone prepared by the oxidation and oximation of an 80:20 ratio of milbemycin A3 and A4; nematocide and insecticide activity via the potentiation of glutamate and GABA-gated chloride-channel opening
Catalog No. 50-297-9917 Supplier Cayman Chemical Supplier No. 1716450 MG
A semisynthetic macrocyclic lactone prepared by the oxidation and oximation of an 80:20 ratio of milbemycin A3 and A4; nematocide and insecticide activity via the potentiation of glutamate and GABA-gated chloride-channel opening