Learn More
Selleck Chemical LLC Milciclib (PHA-848125) 5mg 802539-81-7
SDP
Catalog No. 501948015
Encompass_Preferred
Milciclib (PHA-848125) is a potent, ATP-competitive CDK inhibitor for CDK2 with IC50 of 45 nM. It is >3-fold more selective for CDK2 than CDK1, 2, 4, 5, and 7. Milciclib (PHA-848125) induces cell death through autophagy. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8015 Supplier Selleck Chemical LLC Supplier No. S27515MG
Milciclib (PHA-848125) is a potent, ATP-competitive CDK inhibitor for CDK2 with IC50 of 45 nM. It is >3-fold more selective for CDK2 than CDK1, 2, 4, 5, and 7. Milciclib (PHA-848125) induces cell death through autophagy. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.