Learn More
MSC Ashburn Chemical Technologies Rust Remover, Mild, Neutral pH, 55 gal Drum
Catalog No. 502837646
Encompass
ZAP Rust Remover is a safe and powerful rust and oxidation remover designed to quickly remove rust and corrosion from steel, cast iron, and other ferrous metals. It is safe for nonferrous metals like aluminum, copper, and brass, as well as rubber and plastics. The product features special wetting agents that allow it to quickly penetrate into crevices and metal pores, ensuring thorough rust and corrosion removal.
  • Removes rust and corrosion
  • Safe formula with no harmful acids or caustics
  • Free from harsh fumes
  • Non-flammable
  • pH neutral
  • Water soluble
  • Biodegradable
  • Penetrates quickly into crevices and metal pores
  • Safe for nonferrous metals, rubber, and plastics
Catalog No. 50-283-7646 Supplier MSC Supplier No. 48465892
ZAP Rust Remover is a safe and powerful rust and oxidation remover designed to quickly remove rust and corrosion from steel, cast iron, and other ferrous metals. It is safe for nonferrous metals like aluminum, copper, and brass, as well as rubber and plastics. The product features special wetting agents that allow it to quickly penetrate into crevices and metal pores, ensuring thorough rust and corrosion removal.
  • Removes rust and corrosion
  • Safe formula with no harmful acids or caustics
  • Free from harsh fumes
  • Non-flammable
  • pH neutral
  • Water soluble
  • Biodegradable
  • Penetrates quickly into crevices and metal pores
  • Safe for nonferrous metals, rubber, and plastics