Learn More
MSC MaryKate Mildew Stain Remover, Liquid, Spray Bottle, 32 oz
Catalog No. 502815485
Encompass
MaryKate Mildew Stain Remover is a fast and effective solution designed to clean mildew stains from various marine fabrics and hard surfaces. This water-based formula comes in a fine mist spray bottle, eliminating the need for scrubbing.
  • Fine mist spray won't run, even on vertical surfaces
  • No scrubbing necessary, just spray on, wipe off
  • Use on any vinyl or hard surface
  • Non-flammable
  • Water based
Catalog No. 50-281-5485 Supplier MSC Supplier No. 62194311
MaryKate Mildew Stain Remover is a fast and effective solution designed to clean mildew stains from various marine fabrics and hard surfaces. This water-based formula comes in a fine mist spray bottle, eliminating the need for scrubbing.
  • Fine mist spray won't run, even on vertical surfaces
  • No scrubbing necessary, just spray on, wipe off
  • Use on any vinyl or hard surface
  • Non-flammable
  • Water based