Learn More
Medchemexpress LLC HY-B1836 100mg Medchemexpress, Mildronate CAS:76144-81-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB1836100MG
Encompass_Preferred
Medchemexpress, HY-B1836 100mg Mildronate CAS:76144-81-5 Mildronate (Meldonium) functions as a cardioprotective drug by cpmpetetively inhibiting BBOX1 and OCTN2. Mildronate (Meldonium) exhibits IC50 values of 34-62 μM for human recombinant BBOX and an EC50 of 21 μM for human OCTN2. Mildronate (Meldonium) treatment-induced redirection of long-chain FA metabolism from mitochondria to peroxisomes. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0169