Learn More
Selleck Chemical LLC Mildronate 50mg 86426-17-7 Meldonium, MET-88, Quaterin
SDP
Catalog No. 501363702
Encompass_Preferred
Mildronate (Meldonium, MET-88, Quaterin) is an inhibitor of biosynthesis of L-carnitine by gamma-butyrobetaine (GBB) hydroxylase and as a competitive inhibitor of renal carnitine reabsorption. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3702 Supplier Selleck Chemical LLC Supplier No. S413050MG
Mildronate (Meldonium, MET-88, Quaterin) is an inhibitor of biosynthesis of L-carnitine by gamma-butyrobetaine (GBB) hydroxylase and as a competitive inhibitor of renal carnitine reabsorption. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.