Learn More
AdipoGen Mildronate dihydrate (25 mg)
SDP
Supplier:  AdipoGen CDXM0558M025
Encompass_Preferred
Chemical. CAS: 86426-17-7. Formula: C6H14N2O2 . 2H2O. MW: 182.22. Mildronate is a structural analog of gamma-butyrobetaine (gammaBB), an intermediate in the biosynthesis of carnitine and a modulator of energy metabolism. It blocks carnitine synthesis by inhibiting gammaBB hydroxylase (BBOX) and, less potently, carnitine acetyltransferase (CrAT). Through these actions, mildronate reduces the levels of free carnitine and long chain acyl carnitine. This leads to suppressed fatty acid metabolism and mitochondrial uncoupling during oxidative conditions, resulting in cardioprotective, antidiabetic and neuroprotective effects. Mildronate also improves cognition and reduces amyloid-beta pathology in a mouse model of Alzheimer’s disease.
Catalog No. 50-205-4546