Learn More
Cayman Chemical MIldronatehydrate 10mg
SDP
Catalog No. 502968890
Encompass_Preferred
A structural analog of γBB that inhibits γBB hydroxylase (IC50 = 62 µM) and carnitine acetyltransferase (Ki = 1.6 mM); has cardioprotective and neuroprotective effects; improves cognition and reduces amyloid-β pathology in a mouse model of Alzheimer’s disease
Catalog No. 50-296-8890 Supplier Cayman Chemical Supplier No. 1599710 MG
A structural analog of γBB that inhibits γBB hydroxylase (IC50 = 62 µM) and carnitine acetyltransferase (Ki = 1.6 mM); has cardioprotective and neuroprotective effects; improves cognition and reduces amyloid-β pathology in a mouse model of Alzheimer’s disease