Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Milestone Medical ProWAVE Dehydrating/Clearing Solution
SDP

Catalog No. 22310286 Shop All Milestone Medical Technologies Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
4 x 1 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-310-286 4 x 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-310-286 Supplier Milestone Medical Supplier No. 51410C
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with low-fat specimens

  • Available with or without status color indicator
  • Simple one-step dehydration and clearing
  • Eliminates exposure to and disposal costs of xylene

Specifications

Content And Storage With Color indicator
Product Type Tissue Clearing Reagent
Quantity 4 x 1 gal.
For Use With (Application) Low-fat specimens
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.