Learn More
Bioworld Milieu de Bouturage (S-Medium), 50 L
Catalog No. 50254792
Encompass_Preferred
Milieu de Bouturage (S-Medium)Contains Micronutrients as described by Heller (1953), half concentration of Macronutrients as described by Murashige & Skoog (1962), Vitamins as described by Morel & Wetmore (1951), and SucroseBuffered with MESCytokines and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture tested
Catalog No. 50-254-792 Supplier Bioworld Supplier No. 306301164
Milieu de Bouturage (S-Medium)Contains Micronutrients as described by Heller (1953), half concentration of Macronutrients as described by Murashige & Skoog (1962), Vitamins as described by Morel & Wetmore (1951), and SucroseBuffered with MESCytokines and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture tested