Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Militarine | 58139-23-4 | 99.9% | 5 MG
SDP

Catalog No. 5000231380
Encompass_Preferred

Militarine is a glycosidic natural product isolated from the orchid Bletilla striata, supplied as a solid for research use and reported to exhibit plant growth-inhibitory activity. Typical specifications include CAS 58139-23-4, molecular formula C34H46O17, molecular weight 726.72 g/mol, and high analytical purity.

  • High purity suitable for analytical and biological research.
  • White to light yellow solid.
  • Molecular formula C34H46O17; molecular weight 726.72 g/mol.
  • Storage: 4°C, protect from light; in solvent -80°C (6 months), -20°C (1 month).
  • Reported plant growth-inhibitory activity; intended for research use only.

Catalog No. 50-002-31380 Supplier Medchemexpress LLC Supplier No. HY1223085MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Militarine is a glycosidic natural product isolated from the orchid Bletilla striata, supplied as a solid for research use and reported to exhibit plant growth-inhibitory activity. Typical specifications include CAS 58139-23-4, molecular formula C34H46O17, molecular weight 726.72 g/mol, and high analytical purity.

  • High purity suitable for analytical and biological research.
  • White to light yellow solid.
  • Molecular formula C34H46O17; molecular weight 726.72 g/mol.
  • Storage: 4°C, protect from light; in solvent -80°C (6 months), -20°C (1 month).
  • Reported plant growth-inhibitory activity; intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.