Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSC Go Science Crazy Military Field Compass
SCI_ED

Catalog No. s28113 Shop All GSC Products
Encompass
Change view
Click to view available options
For Use With (Equipment):
Works with USGS topographical maps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
S28113 Works with USGS topographical maps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options

Not a toy. Product intended for ages 13 and up.

Catalog No. S28113 Supplier GSC Go Science Crazy Supplier No. CPFLD10
Only null left
Add to Cart
Add to Cart

Not a toy. Product intended for ages 13 and up.

Accurate and Shock resistant compass preferred by military forces.

  • Contains several features
  • Works with USGS topographical maps
  • Graduated in 6400 mil increments
  • Lens with dial graduated in 0 to 360 degrees in 5 degree increments
  • Calibrated to North America
  • Includes instructions, sighting wire, rotating bezel

Specifications

Description Lens with dial graduated in 0 to 360 degrees in 5 degree increments
For Use With (Equipment) Works with USGS topographical maps
Product Type Field compass
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.