Learn More
Seracare Life Sciences Inc MILK DILUENT/BLOCKING CONC
Supplier:  Seracare Life Sciences Inc 51400011
Encompass
Convenient antibody diluent/blocking solution concentrate containing 2% nonfat milk in a borate buffer. Recommended for blocking membranes in Western blots or as an alternative blocker in microwell ELISA.
Catalog No. 50-674-40