Learn More
Research Products International Corp Dry Powder Milk, 100 Grams
SDP
Catalog No. 50488784
Encompass_Preferred
Nonfat dry milk for use as a blocking agent during Western Blotting to block non-specific antibody binding and reduce background signal.
Catalog No. 50-488-784 Supplier Research Products International Corp Supplier No. M17200100.0
Nonfat dry milk for use as a blocking agent during Western Blotting to block non-specific antibody binding and reduce background signal.