Learn More
NEOGEN Milk Plate Count Agar, 5 kg
SDP
Catalog No. 502015496
Encompass_Preferred
Milk Plate Count Agar is used for the isolation and enumeration of microorganisms in milk and dairy products and is not intended for use in the diagnosis of disease or other conditions in humans.A medium recommended by the British Standards Institute and the International Organization for Standardization for the enumeration of viable bacteria in milk and other dairy products.For more information visit https://www.neogen.com/solutions/Microbiology/milk-plate-count-agar
Catalog No. 50-201-5496 Supplier NEOGEN Supplier No. 700003309
Milk Plate Count Agar is used for the isolation and enumeration of microorganisms in milk and dairy products and is not intended for use in the diagnosis of disease or other conditions in humans.A medium recommended by the British Standards Institute and the International Organization for Standardization for the enumeration of viable bacteria in milk and other dairy products.For more information visit https://www.neogen.com/solutions/Microbiology/milk-plate-count-agar