Learn More
Weber Scientific MILK SMEAR SLIDES
SDP
Catalog No. NC0432995
Encompass
72 slides/pack. Hydrophobic yellow coating delineating the 5 circles on these slides permits rapid smearing of 0.01 ml of milk over an exact 1 cm2 area.
Catalog No. NC0432995 Supplier Weber Scientific Supplier No. 200949
72 slides/pack. Hydrophobic yellow coating delineating the 5 circles on these slides permits rapid smearing of 0.01 ml of milk over an exact 1 cm2 area.