Learn More
Weber Scientific MILK SMEAR SLIDES GRN 72 SLDS
SDP
Catalog No. NC0616007
Encompass
Hydrophobic green coating delineating the 5 circles on these slides permits rapid smearing of 0.01 ml of milk over an exact 1 cm2 area.
Catalog No. NC0616007 Supplier Weber Scientific Supplier No. 200947
Hydrophobic green coating delineating the 5 circles on these slides permits rapid smearing of 0.01 ml of milk over an exact 1 cm2 area.