Learn More
StatLab™ Millennia™ Command Adhesion Microscope Slides
GSA_VASDP
Catalog No. STLMCOMMG90
Change view
Click to view available options
Color:
Aqua
Blue
Green
Lilac
Pink
Red
Tan
White
Yellow
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
STLMCOMMG90 Green
STLMCOMMB90 Blue
STLMCOMMP90 Pink
STLMCOMMY90 Yellow
STLMCOMMW90 White
STLMCOMMA90 Aqua
STLMCOMML90 Lilac
STLMCOMMT90 Tan
STLMCOMMR90 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. STLMCOMMG90 Supplier StatLab™ Supplier No. MCOMM/G/90
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Strong adhesive slides with a hydrophilic surface for use in IHC, special staining and routine histology.
  • Simplify workflow and improve performance with Millennia Command Slides - a true all-purpose slide:
    • A hybrid slide featuring a hydrophilic surface suitable for IHC, H&E and special staining
    • Optimized coating for strong adhesion to reduce tissue loss
    • Consistent staining with minimal background staining during all types of staining applications
    • Compatible with thermal transfer printers
    • Slide features 90-degree corners
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Sterility Non-sterile
Edges 90°, Ground
Material High Quality Glass
Precleaned Yes
Type Adhesion Slide
Frosted 1 End, 1 Side
Color Green