Learn More
MSC Miller Anchor Cable Sling, Steel, Vinyl Coated Wire Rope, 6 ft OAL
Catalog No. 5000509733
Encompass
This anchor cable sling is constructed from steel and vinyl coated wire rope, designed for temporary anchoring applications. It features an O-ring connection and has an overall length of 6 feet.
  • Load capacity of 400 lb.
  • Constructed from steel and vinyl coated wire rope.
  • Silver color.
  • O-ring connection type.
  • Designed for temporary use.
  • Overall length of 6 feet.
  • Overall width of 7-21/64 in.
Catalog No. 50-005-09733 Supplier MSC Supplier No. 59122838
This anchor cable sling is constructed from steel and vinyl coated wire rope, designed for temporary anchoring applications. It features an O-ring connection and has an overall length of 6 feet.
  • Load capacity of 400 lb.
  • Constructed from steel and vinyl coated wire rope.
  • Silver color.
  • O-ring connection type.
  • Designed for temporary use.
  • Overall length of 6 feet.
  • Overall width of 7-21/64 in.