Learn More
Honeywell™ Miller Twin Turbo™ G2 Connector Kit
Catalog No. 19053135
Change view
Click to view available options
Twin Turbo G2 Connector Kit
Catalog No.
19-053-135 Twin Turbo G2 Connector Kit
1 options
Catalog No. 19-053-135 Supplier Honeywell™ Supplier No. MFLC/
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
Includes
Aluminum Carabiner, 3,600 lb. gate strength, G2 Webbing Retainer Clip and G2 D-Pad Connector Clip
For use with Fall Protection Systems
Compliance
Twin Turbo G2 Connector meets all applicable standards including OSHA, ANSI and CSA.
TRUSTED_SUSTAINABILITY
Specifications
Includes Aluminum Carabiner, 3,600 lb. gate strength, G2 Webbing Retainer Clip and G2 D-Pad Connector Clip
Product Type Twin Turbo G2 Connector Kit