Learn More
KLARMANN RULINGS INC Miller Disc Reticle Corner, 21 mm
Catalog No. NC1129244
Encompass_Preferred
A squared reticle can be used for counting. Here the basic principle is that a small area of the specimen is analyzed in order to obtain information about the total area. This removes much irksome work and is preferable in certain instances to analyzing the whole area. For example when it is desired to compare the proportion of large to small particles in a specimen, by using the Miller Disc, the smaller particles may be counted only in the small square, the result being multiplied by ten for comparison with the number of larger particles in the large square.
Catalog No. NC1129244 Supplier KLARMANN RULINGS INC Supplier No. KR80221
A squared reticle can be used for counting. Here the basic principle is that a small area of the specimen is analyzed in order to obtain information about the total area. This removes much irksome work and is preferable in certain instances to analyzing the whole area. For example when it is desired to compare the proportion of large to small particles in a specimen, by using the Miller Disc, the smaller particles may be counted only in the small square, the result being multiplied by ten for comparison with the number of larger particles in the large square.