Learn More
Sklar Instruments Standard Miller Laryngoscope Blade, Sklar Blade Size 00, EA.
SDP
Supplier:  Sklar Instruments 071211
Encompass_Preferred
Standard Miller Laryngoscope Blade, Sklar. Blade Size 00, Sklar Miller Laryngoscope Blades are used for direct, rigid laryngoscopy procedures to view vocal cords, remove foreign objects from the throat, biopsy, or tracheal intubation. The Miller blade is straight and therefore longer than the curved Macintosh blade. It is inserted past the epiglottis, typically causing less trauma during intubation. The Miller blade is commonly used with pediatric and infant patients. This product is a standard, size 00, English Style Miller blade. It has a working length of 43mm and an overall length of 65mm.
Catalog No. 50-118-1301