Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Miller™ Manyard™ Lanyard

Catalog No. 19020533
Change view
Click to view available options
10 ft.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-020533 10 ft.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-020533 Supplier Honeywell™ Supplier No. 216WLS/10FTYL

missing translation for 'validMainframeMsg'

With 20 years of design refinement, the original Manyard Shock-Absorbing Lanyards are unsurpassed in the market for safety, comfort and mobility.

  • Shock-absorbing lanyard greatly reduces fall-arresting force
  • Warning flag pops out when a force of approximately 600 lb. (272kg) has been exerted on the lanyard, indicating that it must not be used again for fall protection
  • Straps are 7/8 in.W (2.2cm) high-visibility nylon with two locking snaps
  • Suitable for use with Type I products
TRUSTED_SUSTAINABILITY

Specifications

Length (English) 10 ft.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.