Learn More
Honeywell™ Miller™ MiniLite™ Self-Retracting Web Lanyard, ANSI A10.32
Catalog No. 19037994
Change view
Click to view available options
Web lanyard
Catalog No.
19-037-994 Web lanyard
1 options
Catalog No. 19-037-994 Supplier Honeywell™ Supplier No. FL112/11FT
Fast-acting brake system limits free fall to just inches
Compact, self-retracting lanyard with a braking system that activates quickly to limit a free fall — by limiting the fall, the risk of injuries is minimized and any necessary rescue is simplified.
Compliance
Meet ANSI™ Z459.1 and ANSI A10.14, CSA Z259.2, CE, and all OSHA requirements.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Web lanyard
Certifications/Compliance ANSI A10.32 Compliant
Length (Metric) 3.3 m
Length (English) 11 ft.