Learn More
Grainger Miller® Air Shield Deflector, 30 to 60 A, Plasma Cutting
Catalog No. 502755400
Encompass_Preferred
This air shield deflector is designed for use with compatible plasma cutting torches to support extended cutting applications. It is engineered to facilitate proper air flow during operation within a current range of 30 A to 60 A.
  • Designed for air-based plasma cutting processes
  • Compatible with handheld torches including XT30, XT30C, XT40, and XT60 series
  • Suited for use with Spectrum 375 and Spectrum 375 X-Treme series plasma cutters
  • Facilitates extended cutting configurations
Catalog No. 50-275-5400 Supplier Grainger Supplier No. 15F418
This air shield deflector is designed for use with compatible plasma cutting torches to support extended cutting applications. It is engineered to facilitate proper air flow during operation within a current range of 30 A to 60 A.
  • Designed for air-based plasma cutting processes
  • Compatible with handheld torches including XT30, XT30C, XT40, and XT60 series
  • Suited for use with Spectrum 375 and Spectrum 375 X-Treme series plasma cutters
  • Facilitates extended cutting configurations