Learn More
Grainger Miller Electric Drag Shield, For XT30 and XT40 Torches
Catalog No. 502757297
Encompass_Preferred
These drag shield caps are engineered for compatibility with specific series of plasma torches, including the XT30, XT30C, XT30R, XT40, XT40R, and XT40M. Designed for handheld plasma cutting processes, these consumables are essential for protecting torch nozzles from contact damage and debris during operation.
  • Protects the plasma torch tip for extended service life
  • Facilitates accurate and easy template cutting
  • Compatible with specific handheld plasma torch series including XT30 and XT40
  • Supports drag shield cutting configurations
Catalog No. 50-275-7297 Supplier Grainger Supplier No. 15F417
These drag shield caps are engineered for compatibility with specific series of plasma torches, including the XT30, XT30C, XT30R, XT40, XT40R, and XT40M. Designed for handheld plasma cutting processes, these consumables are essential for protecting torch nozzles from contact damage and debris during operation.
  • Protects the plasma torch tip for extended service life
  • Facilitates accurate and easy template cutting
  • Compatible with specific handheld plasma torch series including XT30 and XT40
  • Supports drag shield cutting configurations