Learn More
Grainger MILLER Plasma Gouge Shield
Catalog No. 502752743
Encompass_Preferred
The Miller Electric Gouge Shield For Miller XT40 is a replacement shield for the Miller XT40 plasma cutter. It is made of a durable polycarbonate material to protect the operator from sparks and debris when gouging. The shield attaches to the torch using the existing torch screws for easy installation. It measures 5 inches wide by 6 inches high. This shield is an important accessory to protect the operator when performing gouging operations with the XT40 plasma cutter.
Catalog No. 50-275-2743 Supplier Grainger Supplier No. 13Z850
The Miller Electric Gouge Shield For Miller XT40 is a replacement shield for the Miller XT40 plasma cutter. It is made of a durable polycarbonate material to protect the operator from sparks and debris when gouging. The shield attaches to the torch using the existing torch screws for easy installation. It measures 5 inches wide by 6 inches high. This shield is an important accessory to protect the operator when performing gouging operations with the XT40 plasma cutter.