Learn More
Grainger MILLER Remote Control Adapter Cord
Catalog No. 502751741
Encompass_Preferred
The Miller Electric Remote Control Adapter Cord is a 25 foot, 12 gauge, 3 conductor SJOOW cord that allows the user to remotely control a Millermatic or Spectrum plasma cutting system. It has a male plug on one end that connects to the machine and a female receptacle on the other end to plug in the hand held remote. This heavy duty cord is designed for industrial use and features durable rubber insulation to withstand rugged conditions. The remote allows for convenient control of amperage, gas flow, and arc starting up to 25 feet away from the machine. This adapter cord provides an efficient way to remotely operate a welding or cutting system.
Catalog No. 50-275-1741 Supplier Grainger Supplier No. 13Z840
The Miller Electric Remote Control Adapter Cord is a 25 foot, 12 gauge, 3 conductor SJOOW cord that allows the user to remotely control a Millermatic or Spectrum plasma cutting system. It has a male plug on one end that connects to the machine and a female receptacle on the other end to plug in the hand held remote. This heavy duty cord is designed for industrial use and features durable rubber insulation to withstand rugged conditions. The remote allows for convenient control of amperage, gas flow, and arc starting up to 25 feet away from the machine. This adapter cord provides an efficient way to remotely operate a welding or cutting system.