Learn More
Honeywell™ Miller™ Rope Lanyards
Catalog No. 19056756
Change view
Click to view available options
Connector Types:
Lock Snap Loops
Two Locking snaps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connector Types
19-056756 Lock Snap Loops
19-020512 Two Locking snaps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-056756 Supplier Honeywell™ Supplier No. 198RLS2/6FTWH
Rope and web lanyards are recommended for work positioning and restraint only. Length: 6 feet, with snap hook.
Rope lanyards are available in standard nylon or polyester to meet user needs. Standard polyester web lanyards are economical, abrasion resistant and easy to clean. Meets all applicable OSHA, ANSI and CSA standards.
TRUSTED_SUSTAINABILITY
Specifications
Length (English) 6 ft.
Connector Types Lock Snap Loops